Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Bupivacaine Enantiomers on Astec® CHIRALDEX® B-PH
GC Analysis of Bupivacaine Enantiomers on Astec® CHIRALDEX® B-PH
Conditions
column Astec CHIRALDEX B-PH, 10 m x 0.25 mm I.D., 0.12 µm (custom)
carrier gas helium, 12 psi
gradient 130 °C, 2 °C/min to 170 °C
sample peaks 1 & 2: bupivacaine enantiomers
inj. temp. 250 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Enantiomers, Pharmaceuticals
Featured Industry Pharmaceutical (small molecule)
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • Bupivacaine