Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of β-Butyrolactone Enantiomers on Astec® CHIRALDEX® B-DP
GC Analysis of β-Butyrolactone Enantiomers on Astec® CHIRALDEX® B-DP
Conditions
column Astec CHIRALDEX B-DP, 30 m x 0.25 mm I.D., 0.12 µm (78023AST)
oven 140 °C
carrier gas helium, 30 psi
sample peaks 1 & 2: beta-butyrolactone
inj. temp. 250 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Esters, Enantiomers, Lactones
Featured Industry Chemicals and Industrial Polymers
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes