Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of C6-C13 n-Alkanes, BTEX, and Cumene on TCEP (15 m x 0.10 mm I.D., 0.18 μm), Fast GC Analysis
GC Analysis of C6-C13 n-Alkanes, BTEX, and Cumene on TCEP (15 m x 0.10 mm I.D., 0.18 μm), Fast GC Analysis
Conditions
column TCEP, 15 m x 0.10 mm I.D., 0.18 µm (28348-U)
oven 100 °C
carrier gas hydrogen, 40 cm/sec
sample Neat mix, equal volumes of each analyte
injection 0.04 µL, 200:1 split
liner 4 mm I.D., split, cup design
inj. temp. 170 °C
detector FID, 170 °C
Description
Categories Analytical Chromatography, GC Applications, Aromatics, Hydrocarbons
Featured Industry Chemicals and Industrial Polymers
Petroleum