Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Chemical Signals from Spruce Seedlings (Healthy vs. Wounded) on a 5%Phenyl Column after SPME using 100 μm PDMS Fiber
GC Analysis of Chemical Signals from Spruce Seedlings (Healthy vs. Wounded) on a 5%Phenyl Column after SPME using 100 μm PDMS Fiber
Conditions
sample/matrix headspace surrounding Picea abies seedlings
desorption process 225 °C for 30 sec
extraction headspace, 60 min.
SPME fiber 100 µm polydimethylsiloxane (57300-U)
column 5% phenyl polysiloxane, 30 m x 0.25 mm I.D., 0.25 µm (Supelco equivalent, Equity-5, 28089-U)
oven 40 °C (4 min), 4 °C/min to 200 °C (16 min)
carrier gas helium, 10 psi
detector EID, 70ev
Description
Categories Analytical Chromatography, SPME Applications, GC Applications, Agriculture, Volatiles
Featured Industry Agriculture
suitability application for SPME, application for GC