Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Class 1 Residual Solvents on Equity®-1 after SPME using 100 μm PDMS Fiber
GC Analysis of Class 1 Residual Solvents on Equity®-1 after SPME using 100 μm PDMS Fiber
Conditions
sample/matrix 2 mL water + sodium chloride at 25% containing each analyte at 1 ppm, in a 4 mL vial
desorption process 250 °C for 3 min
extraction heated headspace, 50 °C for 5 min
SPME fiber 100 µm PDMS, 24 ga
column Equity-1, 10 m x 0.20 mm I.D., 1.2 µm (28043-U)
oven 40 °C (0.75 min), 20 °C/min to 200 °C (10 min)
carrier gas helium, 35 cm/sec @ 40 °C
injection 5:1 split
liner 0.75 mm I.D. SPME type
inj. temp. 250 °C
detector FID
Description
Categories Analytical Chromatography, SPME Applications, GC Applications, Solvents
Featured Industry Pharmaceutical (small molecule)
Legal Information Equity is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for GC
Analyte
  • 1,1,1-Trichloroethane
  • 1,1-Dichloroethane
  • 1,2-Dichloroethane
  • Benzene
  • Carbon tetrachloride