Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Class 3 Residual Solvents on VOCOL® after SPME using 100 μm PDMS Fiber
GC Analysis of Class 3 Residual Solvents on VOCOL® after SPME using 100 μm PDMS Fiber
Conditions
sample/matrix 2 mL water + sodium chloride at 25% (pH 2) containing each analyte at 5 ppm, in a 4 mL vial
desorption process 250 °C for 3 min
extraction heated headspace, 60 °C for 5 min
SPME fiber 100 µm PDMS, 24 ga, manual (57300-U)
column VOCOL, 10 m x 0.20 mm I.D., 1.2 µm (24129-U)
oven 40 °C (0.75 min), 20 °C/min to 200 °C (10 min)
carrier gas helium, 35 cm/sec @ 40 °C
injection 5:1 split
liner 0.75 mm I.D. SPME type
inj. temp. 250 °C
detector FID
Description
Categories Analytical Chromatography, SPME Applications, GC Applications, Solvents
Featured Industry Pharmaceutical (small molecule)
Legal Information VOCOL is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for GC
Analyte
  • 1-Butanol
  • 1-Propanol
  • 2-Butanone
  • 2-Propanol
  • 4-Methyl-2-pentanone
  • Acetone
  • Anisole
  • Butyl acetate
  • Cumene
  • Dimethyl sulfoxide
  • Ethanol
  • Ethyl acetate
  • Ethyl ether
  • Ethyl formate
  • Heptane
  • Isoamyl alcohol
  • Isobutanol
  • Isobutyl acetate
  • Isopropyl acetate
  • Methyl acetate
  • Methyl-t-butyl ether
  • n-Amyl alcohol
  • Pentane
  • Propyl acetate
  • sec-Butanol
  • Tetrahydrofuran (THF)