GC Analysis of Cold Pressed Bergamot Essential Oil on SLB®-5ms
GC Analysis of Cold Pressed Bergamot Essential Oil on SLB®-5ms
column SLB-5ms, 30 m x 0.25 mm I.D., 0.25 µm (28471-U)
oven 50 °C, 3 °C/min to 250 °C (5 min)
carrier gas helium, 30 cm/sec, constant
sample cold pressed bergamot oil
injection Wet needle, 100:1 split
inj. temp. 250 °C
detector FID, 300 °C
Analysis Note Bergamot oil is used in cosmetic preparations such as perfumes and lotions, and in flavor formulations included in various candies and beverages. The cold-pressed oil is obtained by the rasping of the peel of the citrus bergamia fruit. The oil is commonly treated prior to use to remove furocoumarins and bergapten, which have the potential to cause advserse skin reactions. In addition, terpenes are often reduced or removed to enhance the desirable top-notes of the oil (1). This application illustrates the analysis of a sample of cold-pressed bergamot oil on the SLB-5ms.
Categories Analytical Chromatography, GC Applications, Essential Oils
Featured Industry Flavors and Fragrances
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Gas Purification and Gas Management