Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of D & L-Carvone Enantiomers in Caraway Essential Oil (Spiked with L-Carvone Enantiomer) on Astec® CHIRALDEX® G-TA
GC Analysis of D & L-Carvone Enantiomers in Caraway Essential Oil (Spiked with L-Carvone Enantiomer) on Astec® CHIRALDEX® G-TA
Conditions
column CHIRALDEX G-TA, 30 m x 0.25 mm I.D., 0.12 µm (73033AST)
oven 40 °C (1 min), then 2 °C/min to 170 °C (15 min)
carrier gas helium, 25cm/ sec set at 110C
sample Dill Seed Oil + L- Carvone Spike, neat
injection 0.1 µL100:1 split
liner 4 mm I.D. cup injector
inj. temp. 250 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Essential Oils, Enantiomers, Ketones
Featured Industry Flavors and Fragrances
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes