GC Analysis of Dibasic Acid Methyl Esters on SP™-2380
GC Analysis of Dibasic Acid Methyl Esters on SP™-2380
column SP-2380, 30 m x 0.25 mm I.D., 0.20 µm (24110-U)
oven 170 °C, 4 °C/min to 200 °C
carrier gas helium, 20 cm/sec @ 185 °C
sample 1000 µg/mL each analyte in methylene chloride
injection 0.1µL, split 100:1
inj. temp. 250 °C
detector FID, 250 °C
Categories Analytical Chromatography, GC Applications, Fatty Acid Methyl Esters (FAMEs)
Featured Industry Chemicals and Industrial Polymers
Legal Information SP is a trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
  • Dimethyl adipate
  • Dimethyl azelate
  • Dimethyl glutarate
  • Dimethyl isophthalate
  • Dimethyl malonate
  • Dimethyl pimelate
  • Dimethyl sebacate
  • Dimethyl suberate
  • Dimethyl succinate
  • Dimethyl terephthalate
  • Dimethylphthalate
Related Links