Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of FAMEs in B20 Biodiesel (Soy Source) on Omegawax®
GC Analysis of FAMEs in B20 Biodiesel (Soy Source) on Omegawax®
Conditions
column Omegawax 250, 30 m x 0.25 mm I.D., 0.25 µm (24136)
oven 50 °C, 13 °C/min to 270 °C (5 min)
sample B20 biodiesel (soy source) diluted 1:20 in n-hexane
injection 1 µL100:1 split
liner 4 mm I.D. FocusLiner™ inlet liner (no taper)
inj. temp. 250 °C
detector helium, 40 cm/sec
detector FID, 270 °C
Description
Categories Analytical Chromatography, GC Applications, Hydrocarbons, Fatty Acid Methyl Esters (FAMEs)
Featured Industry Biofuels
Legal Information Omegawax is a registered trademark of Sigma-Aldrich Co. LLC, FocusLiner is a trademark of SGE Analytical Science Pty Ltd.
Other Notes Complementary products:
Syringes