Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of FAMEs in Cod Liver Oil on Omegawax®
GC Analysis of FAMEs in Cod Liver Oil on Omegawax®
Conditions
column Omegawax 250, 30 m x 0.25 mm I.D., 0.25 µm (24136)
oven 180 °C, 3 °C/min to 270 °C (2 min)
carrier gas hydrogen, 35 cm/sec constant
sample cod liver oil FAMEs in hexane
injection 0.2 µL in the split mode (50:1)
inj. temp. 280 °C
detector FID, 280 °C
Description
Categories Analytical Chromatography, GC Applications, Fatty Acid Methyl Esters (FAMEs)
Featured Industry Food and Beverages
Legal Information Omegawax is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes