Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of FAMEs in Cookies on SP™-2560 after SPE Fractionation using Discovery® Ag-Ion
GC Analysis of FAMEs in Cookies on SP™-2560 after SPE Fractionation using Discovery® Ag-Ion
Conditions
sample preparation SPE (Solid Phase Extraction)
sample/matrix 1 g of commercially purchased cookies was ground and subjected to acid digestion and alkali hydrolysis followed by methylation as described in AOCS Official Method Ce 1k-09
SPE tube/cartridge Discovery Ag-Ion SPE tubes, 750 mg/6 mL (54225-U)
condition 4 mL of acetone; allow solvent to gravity drip completely through tube; discard eluant; 4 mL of hexane; allow solvent to gravity drip completely through tube; discard eluant
sample addition 1 mL of extract; discard any eluant that drips through tube
elution (Fraction 1) 6 mL of hexane:acetone (96:4); collect eluant in a fresh container with slight vacuum; ((Fraction 2) 4 mL of hexane: acetone (90:10); collect eluant in a fresh container with slight vacuum;(Fraction 3) 4 mL of 100% acetone; collect eluant in a fresh container with slight vacuum)
eluate post-treatment evaporate each fraction at room temperature using nitrogen; reconstitute each fraction to 1 mL of hexane
column SP-2560, 100 m x 0.25 mm I.D., 0.20 µm (24056)
oven 180 °C
carrier gas hydrogen, 1 mL/min
injection 1 µL, 10:1 split
liner 4 mm I.D., split type, single taper wool packed FocusLiner™ design
inj. temp. 250 °C
detector FID, 250 °C
Description
Analysis Note A sample of commercially purchased cookies were prepared using an acid digestions /alkali hydrolysis followed by methylation as described in AOCS Official Method Ce 1k-09 (1). The resulting extracts were then fractionated using Ag-ion SPE and analyzed by GC/FID on the 100 m SP-2560 °Capillary column.
Categories Analytical Chromatography, SPE Applications, GC Applications, FAMEs (Fatty Acid Methyl Esters), Fatty Acid Methyl Esters (FAMEs)
Featured Industry Food and Beverages
Legal Information FocusLiner is a trademark of SGE Analytical Science Pty Ltd., Discovery is a registered trademark of Sigma-Aldrich Co. LLC, SP is a trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC, application for SPE