GC Analysis of FAMEs in Cow's Milk on SLB®-IL111
GC Analysis of FAMEs in Cow's Milk on SLB®-IL111
column SLB-IL111, 100 m x 0.25 mm I.D., 0.20 µm (29647-U)
oven 50 ºC (4 min), 8 ºC/min to 160 °C (25 min), 4 ºC/min to 220 ºC
carrier gas hydrogen, 40 cm/sec
sample FAMEs from 100 mg cow milk transesterified in sodium methoxide/methanol + boron trifluoride
injection 0.2 µL10:1 split
inj. temp. 230 °C
detector FID, 240 ºC
Categories Analytical Chromatography, Food & Beverage Analysis, GC Applications, Beverage Testing, Milk, Fatty Acid Methyl Esters (FAMEs)
Featured Industry Food and Beverages
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Gas Purification and Gas Management