Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of FAMEs in Rapeseed Oil (with Conjugated Linoleic Acid (CLA) FAME Isomers Added) on SP™-2560
GC Analysis of FAMEs in Rapeseed Oil (with Conjugated Linoleic Acid (CLA) FAME Isomers Added) on SP™-2560
Conditions
column SP-2560, 200 m x 0.25 mm I.D., 0.20 µm (29688-U)
oven 175 °C
carrier gas hydrogen
sample Mix of rapeseed oil FAME isomers and CLA FAME isomers
injection 1µL, split 50:1
liner 4 mm I.D., split type, cup design
inj. temp. 250 °C
detector FID, 250 °C
Description
Analysis Note Four key conjugated linoleic acid (CLA) FAME isomers are resolved in a mixture with rapeseed oil FAME isomers.
Categories Analytical Chromatography, GC Applications, Fatty Acid Methyl Esters (FAMEs)
Featured Industry Agriculture
Food and Beverages
Legal Information SP is a trademark of Sigma-Aldrich Co. LLC