GC Analysis of Fuel Oil #2 on Equity®-1
GC Analysis of Fuel Oil #2 on Equity®-1
column Equity-1, 30 m x 0.25 mm I.D., 0.25mum (28046-U)
oven 40 °C (4 min), 8 °C/min to 325 °C (5 min)
carrier gas helium, 30 cm/sec constant
sample 100 ng on-column of a No. 2 Fuel Oil standard (47515-U)
injection 1.0 µL, 200:1 split
liner 4 mm I.D., single taper
inj. temp. 275 °C
detector FID, 325 °C
Categories Analytical Chromatography, GC Applications, Hydrocarbons
Featured Industry Petroleum
Legal Information Equity is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Gas Purification and Gas Management