Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Fuel Oil #3 on Equity®-1 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
GC Analysis of Fuel Oil #3 on Equity®-1 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Conditions
column Equity-1, 15 m x 0.10 mm I.D., 0.10 µm (28039-U)
oven 80 °C, 50 °C/min to 325 °C
carrier gas hydrogen, 45 cm/sec constant
sample No.3 Fuel Oil standard, 50 mg/mL in hexane (47534-U)
injection 0.3 µL, 200:1 split, 0.02 min pre-injection dwell time
liner 2 mm I.D., straight
inj. temp. 250 °C
detector FID, 350 °C
Description
Categories Analytical Chromatography, GC Applications, Hydrocarbons
Featured Industry Petroleum
Legal Information Equity is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes