Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Glycerides in Lard on MET-Biodiesel
GC Analysis of Glycerides in Lard on MET-Biodiesel
Conditions
column MET-Biodiesel, 14 m x 0.53 mm I.D., 0.16 µm with integrated 2 m x 0.53 mm I.D. guard (28668-U)
oven 150 °C, 20 °C/min to 350 °C (15 min)
carrier gas helium, 15 mL/min
sample lard extract
injection 1 µL, cool on-column
detector FID, 400 °C
Description
Categories Analytical Chromatography, GC Applications, Glycerides
Featured Industry Food and Beverages
Other Notes Complementary products:
Syringes