GC Analysis of Glycols on Nukol™
GC Analysis of Glycols on Nukol™
column Nukol, 15 m x 0.53 mm I.D., 0.5 µm (25326)
oven 110 °C, 8 °C/min to 220 °C
carrier gas helium, 15 mL/min
sample 100 ppm each glycol in water
injection 1 µL, direct
detector FID, 250 °C
Categories Analytical Chromatography, GC Applications, Diols, Glycols
Featured Industry Chemicals and Industrial Polymers
Cosmetics, Personal Care, and Cleaning Products
Food and Beverages
Legal Information Nukol is a trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
  • 1,2-Propanediol
  • 1,4-Butanediol
  • 2,3-Butanediol
  • Diethylene glycol
  • Dipropylene glycol
  • Ethylene glycol
  • Glycerol
  • Tetraethylene glycol
  • Triethylene glycol
Related Links