GC Analysis of Grob Test Mix on SLB®-IL76i
GC Analysis of Grob Test Mix on SLB®-IL76i
column SLB-IL76i, 30 m x 0.25 mm I.D., 0.20 µm (29869-U)
oven 55 °C (2 min), 2 °C/min to 180 °C (10 min)
carrier gas helium, 1.2 mL/min (28 cm/sec)
sample programmed test mix (47304), 12 analytes at varied concentrations in methylene chloride
injection 1 µL, 100:1 split
liner 4 mm I.D., split type, cup design
inj. temp. 250 °C
detector FID, 250 °C
Analysis Note The i-series of ionic liquid capillary GC columns are characterized by their improved inertness for polar analytes. This application shows that the SLB IL76i can generate great peak shapes for analytes with various functionality.
Featured Industry General Analytical
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC