Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Impurities in Propylene on Alumina Chloride PLOT
GC Analysis of Impurities in Propylene on Alumina Chloride PLOT
Conditions
column Alumina chloride PLOT, 50 m x 0.53 mm I.D. (28329-U)
oven 35 °C (2.5 min), 5 °C/min to 150 °C (4.5 min)
carrier gas helium, 3.0 mL/min
sample 18-component C1-C5 hydrocarbon standard in nitrogen, spiked into propylene
injection 10 µL, direct using a sampling valve, 150 °C
inj. temp. 200 °C
detector FID, 230 °C
Description
Categories Analytical Chromatography, GC Applications, Light Hydrocarbons, Hydrocarbons
Featured Industry Chemicals and Industrial Polymers
suitability application for GC
Analyte
  • 1,3-Butadiene
  • 1-Butene
  • Acetylene
  • cis-2-Butene
  • Cyclopropane
  • Ethane
  • Ethylene
  • Isobutane
  • Isobutylene
  • Isopentane
  • Methane
  • n-Butane
  • n-Pentane
  • Propadiene
  • Propane
  • Propylene
  • Propyne
  • trans-2-Butene