Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Industrial Solvents on Equity®-1
GC Analysis of Industrial Solvents on Equity®-1
Conditions
column Equity-1, 30 m x 0.32 mm I.D., 1.0 µm (28057-U)
column temp. 35 C (8 min), 4 C/min to 130 C (2 min)
carrier gas helium, 25 cm/sec @ 35 °C
sample 59 component neat solvent mixture
injection 0.5 µL, 200:1split
liner split, cup design
inj. temp. 250 C
detector FID, 250 C
Description
Categories Analytical Chromatography, GC Applications, Solvents
Featured Industry Chemicals and Industrial Polymers
Legal Information Equity is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 1,1,1-Trichloroethane
  • 1,1,2,2-Tetrachloroethane
  • 1,1-Dichloroethane
  • 1,1-Dichloroethylene
  • 1,2-Dichlorobenzene
  • 1,2-Dichloroethane
  • 1,4-Dioxane
  • 1-Propanol
  • 2-Butanone
  • 2-Ethoxyethanol
  • 2-Ethoxyethyl acetate
  • 2-Hexanone
  • 2-Methoxyethanol
  • 2-Methylcyclohexanol
  • 2-Methylphenol
  • 2-Nitropropane
  • 2-Propanol
  • 3-Methylphenol
  • 4-Methyl-2-pentanone
  • 4-Methylphenol
  • 5-Methyl-2-hexanone
  • Acetone
  • Benzene
  • Butyl Cellosolve
  • Carbon tetrachloride
  • Chlorobenzene
  • Chloroform
  • Cyclohexanol
  • Diacetone alcohol
  • Dimethylformamide (DMF)
  • Ethanol
  • Ethyl acetate
  • Ethyl benzene
  • Ethyl formate
  • Hexane
  • Isoamyl acetate
  • Isoamyl alcohol
  • Isobutanol
  • Isobutyl acetate
  • Isopropyl acetate
  • Mesityl oxide
  • Methanol
  • Methyl acetate
  • Methyl formate
  • Methylene chloride
  • m-Xylene
  • n-Amyl acetate
  • n-Butanol
  • n-Butyl acetate
  • n-Propyl acetate
  • o-Xylene
  • p-Xylene
  • sec-Butanol
  • Styrene
  • Tetrachloroethene
  • Tetrahydrofuran (THF)
  • Toluene
  • trans-1,2-Dichloroethylene
  • Trichloroethylene