Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Kerosene on SLB®-5ms (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
GC Analysis of Kerosene on SLB®-5ms (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Conditions
column SLB-5ms, 10 m x 0.10 mm I.D., 0.10 µm (28465-U)
oven 40 °C, 80.0 °C/min to 150 °C, 70.0 °C/min to 250 °C, 50.0 °C/min to 320 °C
carrier gas hydrogen, 85 cm/sec constant
sample kerosene
injection 0.2 µL, 800:1 split
inj. temp. 320 °C
detector FID, 320 °C
Description
Categories Analytical Chromatography, GC Applications, Hydrocarbons
Featured Industry Petroleum
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • Kerosene