GC Analysis of Lactone Enantiomers on β-DEX™ 110
GC Analysis of Lactone Enantiomers on β-DEX™ 110
column beta-DEX 110, 30 m x 0.25 mm I.D., 0.25 µm (24301)
oven 90 °C, 1 °C/min to 200 °C
carrier gas helium, 20 cm/sec @ 90 °C
sample 0.5 mg/mL in methylene chloride
injection 1 µL, 100:1 split
inj. temp. 300 °C
detector FID, 300 °C
Categories Analytical Chromatography, GC Applications, Esters, Enantiomers, Lactones
Featured Industry Chemicals and Industrial Polymers
Flavors and Fragrances
Legal Information DEX is a trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Gas Purification and Gas Management