Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Methylphenidate (Ritalin) Enantiomers (N-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX® B-DA
GC Analysis of Methylphenidate (Ritalin) Enantiomers (N-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX® B-DA
Conditions
column Astec CHIRALDEX B-DA, 30 m x 0.25 mm I.D., 0.12 µm (72023AST)
oven 170 °C
carrier gas helium, 35 psi
sample peaks 1 & 2: methylphenidate enantiomers (N-TFA derivative)
inj. temp. 250 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Chiral Applications, Clinical, Forensic & Toxicology, Enantiomers, Pharmaceuticals, Therapeutic Drug Monitoring, Drugs of Abuse
Featured Industry Clinical
Pharmaceutical (small molecule)
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes