GC Analysis of Organic Acids on Nukol™
GC Analysis of Organic Acids on Nukol™
column Nukol, 15 m x 0.32 mm I.D., 0.25 µm (24130)
oven 80 °C (1 min), 15 °C/min to 200 °C (3 min)
carrier gas helium, 2 mL/min constant
sample 5 analytes, at concentrations indicated in 1 M Phosphoric acid
injection 1 µL100:1 split
liner 4 mm I.D., split, cup design
inj. temp. 250 °C
detector FID, 250 °C
Categories Analytical Chromatography, GC Applications, Fatty Acids, Organic Acids, Acids
Featured Industry Food and Beverages
Legal Information Nukol is a trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Gas Purification and Gas Management