Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Permanent Gases and Light Hydrocarbons on Carboxen-1006 PLOT and a Packed Column (Carboxen® 1004)
GC Analysis of Permanent Gases and Light Hydrocarbons on Carboxen-1006 PLOT and a Packed Column (Carboxen® 1004)
Conditions
column Carboxen 1004 micropacked, 2 m x 1/16 in. stainless steel (12846)
column Carboxen-1006 PLOT, 30 m x 0.53 mm I.D. (25461)
oven 35 °C (1 min), 24 °C/min to 225 °C (micropacked)
oven 35 °C (5 min), 24 °C/min to 225 °C (PLOT)
carrier gas helium, 22 cm/sec
sample 1% each analyte in nitrogen
injection 35 µL
detector TCD, 230 °C
Description
Categories Analytical Chromatography, GC Applications, Volatiles, Light Hydrocarbons, Gases, Hydrocarbons
Featured Industry Chemicals and Industrial Polymers
Legal Information Carboxen is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • Acetylene
  • Carbon dioxide
  • Carbon monoxide
  • Ethane
  • Ethylene
  • Methane
  • Nitrogen