Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Pesticides in Spinach on Equity®-1 after SPE using Supelclean™ ENVI-Carb-II/PSA
GC Analysis of Pesticides in Spinach on Equity®-1 after SPE using Supelclean™ ENVI-Carb-II/PSA
Conditions
sample preparation SPE (Solid Phase Extraction)
sample/matrix 10 g of fresh spinach spiked at 0.2 ppm each pesticide
SPE tube/cartridge Supelclean ENVI-Carb-II/PSA, 500mg/300mg/6mL (55119-U)
condition 5 mL acetonitrile:toluene (3:1, v/v)
sample addition 1 mL concentrated sample extract
elution 20 mL acetonitrile:toluene (3:1, v/v)
eluate post-treatment Evaporate eluate to dryness with nitrogen at 40 °C. Reconstitute in 1 mL acetone:hexane (1:1, v/v).
column Equity-1, 30 m x 0.25 mm I.D., 0.25 µm (28046-U)
oven 50 °C (5 min), 25 °C/min to 125 °C, 10 °C/min to 30 °C (8 min)
carrier gas helium, 0.9 mL/min., constant flow mode
injection 1 µL, splitless (splitter open at 1 min)
liner 4 mm I.D., single taper
inj. temp. 200 °C, aux: 325 °C (8 min)
detector MSD, scan range 45-450 amu
Description
Application Sample Pre-treatment:
Homogenize 10 g of spinach in 10 mL of acetonitrile.
Categories Analytical Chromatography, SPE Applications, GC Applications, Food & Beverage Analysis, Pesticides, Food Industry Applications, Pesticide and Metabolite Residues, Applications (Chromatograms)
Featured Industry Agriculture
Food and Beverages
Legal Information Supelclean is a trademark of Sigma-Aldrich Co. LLC, Equity is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC, application for SPE