Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Petitgrain Essential Oil on SLB®-5ms, 50 °C Initial Oven Temp., 30 cm/s
GC Analysis of Petitgrain Essential Oil on SLB®-5ms, 50 °C Initial Oven Temp., 30 cm/s
Conditions
column F.S. CAP. SLB-5ms 30 m x 0.25 mm I.D., 0.25 µm (58471-U)
oven 50 °C, 3 °C/min to 250 °C (10 min)
carrier gas helium, 30.0 cm/s, 99.5 kPa
sample (oil obtained by steam distillation of the leaves and twigs, diluted)
injection 1.0 µL, 100:1 split
inj. temp. 250 °C
detector FID, 280 °C
Description
Analysis Note The bitter orange tree is used to obtain different products: essential oil cold pressed from the peel of the fruit, the neroli distilled from the flowers, the petitgrain distilled from the leaves and twigs, and different other products obtained by solvent extraction or by hydrodistillation. All of these products are much appreciated for their odor properties. Their analytical characterization is important for the identification of quality parameters, and considering their commercial value, it is important to determine their composition. This study is meant to determine differences between Italian and Egyptian cold-pressed, neroli and petitgrain oils, and gives useful information on the composition of the essential oil obtained from the whole crushed fruits. The composition of all the samples was studied by different analytical techniques to determine the volatile fraction, the enantiomeric distribution of some volatiles (by GC, GC/MS and esGC, respectively) and the oxygen heterocyclic fraction by RP-HPLC.
Categories Analytical Chromatography, GC Applications, Essential Oils
Featured Industry Flavors and Fragrances
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • (E)-2-Hexenal
  • (E)-2-Hexenyl acetate
  • (E)-Nerolidol
  • (E)-β-Farnesene
  • (E)-β-Ocimene
  • (E,E)-α-Farnesene
  • (Z)-2-Hexenol
  • (Z)-3-Hexenol
  • (Z)-3-Hexenyl acetate
  • (Z)-β-Ocimene
  • 1,8-Cineole
  • 6-Methyl-5-hepten-2-one
  • Bicyclogermacrene
  • Camphene
  • Caryophyllene oxide
  • cis-Linalool oxide
  • cis-p-Menth-2-en-1-ol
  • Citronellal
  • Geranial
  • Geranic acid methyl ester
  • Geraniol
  • Geranyl acetate
  • Hexanol
  • Limonene
  • Linalool
  • Linalyl acetate
  • Linalyl propionate
  • Methyl-N-methyl anthranilate
  • Myrcene
  • Neral
  • Nerol
  • Neryl acetate
  • p-Cymen-8-ol
  • p-Cymene
  • Sabinene
  • Spathulenol
  • Terpinen-4-ol
  • Terpinolene
  • α-Huµlene
  • α-Phellandrene
  • α-Pinene
  • α-Terpinene
  • α-Terpineol
  • α-Terpinyl acetate
  • α-Thujene
  • β-Caryophyllene
  • β-Elemene
  • β-Pinene
  • β-Sesquiphellandrene
  • γ-Terpinene
  • δ-3-Carene
  • δ-Cadinene
  • δ-Elemene