Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) on Equity®-5 (15 m x 0.10 mm I.D., 0.10 μm), Hydrogen Carrier Gas, Fast GC Analysis
GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) on Equity®-5 (15 m x 0.10 mm I.D., 0.10 μm), Hydrogen Carrier Gas, Fast GC Analysis
Conditions
column Equity-5, 15 m x 0.10 mm I.D., 0.10 µm (28083-U)
oven 100 °C (1 min), 35 °C/min to 325 °C (5 min)
carrier gas hydrogen, 45 cm/sec constant
sample Each analyte at 200 ppm in methylene chloride
injection 1 µL200:1 split
liner 4 mm I.D., split, cup design
inj. temp. 250 °C
detector FID, 350 °C
Description
Categories Analytical Chromatography, GC Applications, PAHs, Hydrocarbons
Featured Industry Environmental
Food and Beverages
Legal Information Equity is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes