GC Analysis of Primary Amines on Carbowax® Amine
GC Analysis of Primary Amines on Carbowax® Amine
column Carbowax Amine, 30m x 0.53 mm I.D., 1.0 µm (25353)
oven 50 °C, 8 °C/min to 200 °C (10 min)
carrier gas helium, 20 cm/sec @ 170 °C
sample neat amines mix
injection 0.02 µL100:1 split
inj. temp. 220 °C
detector FID, 220 °C
Categories Analytical Chromatography, GC Applications, Amines
Featured Industry Chemicals and Industrial Polymers
Legal Information Carbowax is a registered trademark of Union Carbide
Other Notes Complementary products:
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
  • Butylamine
  • Decylamine
  • Heptylamine
  • Hexylamine
  • Nonylamine
  • Octylamine
  • Pentylamine
  • Propylamine
Related Links