Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of PUFA No. 3 Mix (Menhaden Oil Polyunsaturated Fatty Acid (PUFA) Methyl Esters) on SLB®-IL60
GC Analysis of PUFA No. 3 Mix (Menhaden Oil Polyunsaturated Fatty Acid (PUFA) Methyl Esters) on SLB®-IL60
Conditions
column SLB-IL60, 30 m x 0.25 mm I.D., 0.20 µm (29505-U)
oven 170 °C, 1 °C/min to 225 °C
carrier gas helium, 1.2 mL/min
sample PUFA No. 3 Mix (47085-U), diluted in 1 mL of hexane
injection 1.0 µL100:1 split
liner 4 mm I.D., split/splitless type, single taper wool packed FocusLiner™ design
inj. temp. 250 °C
detector FID, 260 °C
Description
Categories Analytical Chromatography, GC Applications, Fatty Acid Methyl Esters (FAMEs)
Featured Industry Agriculture
Food and Beverages
Legal Information FocusLiner is a trademark of SGE Analytical Science Pty Ltd., SLB is a registered trademark of Sigma-Aldrich Co. LLC