Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Resveratrol in Red Wine on SLB®-5ms after SPME using 85 μm Polyacrylate Fiber
GC Analysis of Resveratrol in Red Wine on SLB®-5ms after SPME using 85 μm Polyacrylate Fiber
Conditions
sample/matrix 3 mL of red wine (California merlot) diluted 3:1 in 12% ethanol:water
desorption process 280 °Cfor 2 min
extraction immersion for 15 min at room temperature while stirring at 400 rpm, fiber gently blotted with a Kimwipe to remove excess water, (then derivatization by immersion in a 4 mL vial containing 5 µL of Sylon-BFT (fresh vial for each analysis than was allowed to equilabrate 60-90 minutes prior to use) for 20 min)
SPME fiber 85 µm polyacrylate FS 24gauge manual (57304)
SPME fiber 85 µm polyacrylate FS 23 gauge auto (57294-U)
SPME fiber 85 µm polyacrylate FS 24gauge auto (57305)
column SLB-5ms; 30 m x 0.25 mm I.D., 0.25 µm (28471-U)
oven 100 °C (1 min), 10 °C/min to 325 °C (3 min)
carrier gas helium, 1 mL/min, constant
liner 0.75 mm I.D., SPME
MSD interface 325 °C
scan range m/z 40-450
Description
Categories Analytical Chromatography, Food & Beverage Analysis, SPME Applications, GC Applications, Foods, Flavors & Fragrances, Beverage Testing, Antioxidants, Beer, Wine, and Spirits, Wine
Featured Industry Food and Beverages
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for SPME, application for GC