Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Solvents on a Packed Column (SP-1000 on Carbopack™ C)
GC Analysis of Solvents on a Packed Column (SP-1000 on Carbopack™ C)
Conditions
column 10 ft (3.048m) x 1/8 in.(9.82 mm), stainless steel (11820)
packing Carbopack™ C/SP-1000 (11820)
column temp. A & C - 70 °C to 225 °C at 4 °C/min B - 100 °C to 225 °C at 8 °C/min
carrier gas nitrogen, 20 mL/min
inj. temp. 0.3 µL3% each component by volume (approx. 12 mug each)
detector FID
Description
Categories Analytical Chromatography, GC Applications, Solvents
Featured Industry Chemicals and Industrial Polymers
Legal Information Carbopack is a trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes