GC Analysis of Solvents on PAG
GC Analysis of Solvents on PAG
column PAG, 30 m x 0.25 mm I.D., 0.25 µm (24223)
oven 30 °C (4 min), 4 °C/min to 200 °C (4 min)
carrier gas helium, 20 cm/sec (set at 165 °C)
sample neat solvents mix
injection 0.1 µL, split 100:1
inj. temp. 250 °C
detector FID, 260 °C
Categories Analytical Chromatography, GC Applications, Solvents
Featured Industry Chemicals and Industrial Polymers
Other Notes Complementary products:
Gas Purification and Gas Management