GC Analysis of Trichloroanisole (TCA) and Precursors on SLB®-5ms after SPME using 100 μm a PDMS Fiber
GC Analysis of Trichloroanisole (TCA) and Precursors on SLB®-5ms after SPME using 100 μm a PDMS Fiber
sample/matrix each analyte at 100 ng/L in a 12% ethanol solution
desorption process 250 °C for 3 min
extraction headspace, 50 °C for 30 min, with stirring
SPME fiber metal fiber coated with 100 µm PDMS (57928-U)
column SLB-5ms, 30 m x 0.25 mm I.D., 0.25 µm (28471-U)
oven 50 °C (1 min), 25 °C/min to 280 °C
carrier gas helium, 1.5 mL/min, constant
sample Wine cork
injection splitless/split, closed 2 min
liner 0.75 mm I.D. SPME
inj. temp. 250 °C
detector GC/MS, ECD, 290 °C
Analysis Note 2,4,6-Trichloroanisole (TCA) is a phenolic compound that, if present, gives wine an unpleasant odor often described as musty, woody, dank, and acrid. Human sensory thresholds for TCA are very low, estimated to be approximately 5 parts per trillion. Natural phenolic compounds in cork can convert into TCA when the cork is exposed to chlorine-containing bleach. TCA contamination can also derive from fungi growing on the grapes during fermentation. In this application, a 100 μm PDMS fiber was used to extract TCA from the headspace above a spiked wine sample prior to analysis by GC. The SLB-5ms GC columns provide consistently low-bleed, inert, efficient, and durable separations.
Categories Analytical Chromatography, Food & Beverage Analysis, SPME Applications, GC Applications, Foods, Flavors & Fragrances, Beverage Testing, Volatiles, Beer, Wine, and Spirits, Wine
Featured Industry Food and Beverages
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for SPME, application for GC
Related Links