Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Trichloroanisole (TCA) and Precursors in Red Wine (Spiked) on SLB-5ms after SPME using 100 μm PDMS Fiber
GC Analysis of Trichloroanisole (TCA) and Precursors in Red Wine (Spiked) on SLB-5ms after SPME using 100 μm PDMS Fiber
Conditions
sample/matrix 1.5 mL red wine spiked with each analyte at 100 ng/L + 600 µL 5% potassium carbonate (K2CO3) + 240 mg sodium chloride (NaCl) + 60 µL acetic anhydride
desorption process 250 °C for 3 min
extraction headspace, 50 °C for 30 min, with stirring
SPME fiber metal fiber coated with 100 µm PDMS (57928-U)
column SLB-5ms, 30 m x 0.25 mm I.D., 0.25 µm (28471-U)
oven 50 °C (1 min), 25 °C/min to 280 °C
carrier gas helium, 1.5 mL/min, constant
liner 0.75 mm I.D. SPME
detector ECD, 290 °C
Description
Categories Analytical Chromatography, Food & Beverage Analysis, SPME Applications, GC Applications, Foods, Flavors & Fragrances, Beverage Testing, Volatiles, Beer, Wine, and Spirits, Wine
Featured Industry Food and Beverages
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for SPME, application for GC