Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Volatiles in Kentucky Bourbon (Sample 1) on VOCOL® after SPME using 50/30 μm DVB/Carboxen®/PDMS Fiber, 15%(w/v) Salt Added
GC Analysis of Volatiles in Kentucky Bourbon (Sample 1) on VOCOL® after SPME using 50/30 μm DVB/Carboxen®/PDMS Fiber, 15%(w/v) Salt Added
Conditions
sample preparation Solid Phase Microextraction
sample/matrix 4 mL Kentucky bourbon (sample 1, 80 proof) in a 10 mL vial plus 15% sodium chloride (w/v)
desorption process 270 °C for 3 min
extraction headspace, 40 °C for 15 min with agitation
SPME fiber 50/30 µm divinylbenzene/Carboxen®/polydimethylsiloxane on StableFlex™ (57298-U)
column VOCOL®, 30 m x 0.25 mm I.D., 1.50 µm (24205-U)
oven 40 °C (2.5 min), 6 C/min to 55 °C, 45 ºC/min to 235 °C (5min)
carrier gas helium, 1.0 mL/min
injection splitless (closed 0.75 min)
liner 0.75 mm I.D., direct (SPME) type, straight design
inj. temp. 270 °C
detector FID, 300 °C
Description
Analysis Note Beverages are liquid, so sample preparation is often simple, and can be accomplished with direct injection, headspace, solid phase microextraction (SPME), solid phase extraction (SPE), or liquid-liquid extraction (LLE). SPME coupled with GC is an effective technique for monitoiring the volatiles present in the headspace over spirits.
Categories Analytical Chromatography, SPME Applications, GC Applications, Food and Beverage, Alcohols, Volatiles
Featured Industry Food and Beverages
Legal Information StableFlex is a trademark of Sigma-Aldrich Co. LLC, Carboxen is a registered trademark of Sigma-Aldrich Co. LLC, VOCOL is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for SPME, application for GC