Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Volatiles in Water on VOCOL after SPME using 100 μm PDMS Fiber
GC Analysis of Volatiles in Water on VOCOL after SPME using 100 μm PDMS Fiber
Conditions
sample/matrix 50ppb each analyte in 1.8 mL saturated salt water
desorption process 230 °C, splitless (closed 3 min)
extraction 5 min immersed in water with rapid stirring
SPME fiber polydimethylsiloxane, 100 µm film (57300-U)
column VOCOL, 10 m x 0.20 mm I.D., 1.2 µm (24129-U)
oven 40 °C (0.75 min), 20 °C/min to 160 °C
carrier gas helium, 40 cm/sec
detector FID, 260 °C
Description
Categories Analytical Chromatography, SPME Applications, GC Applications, Foods, Flavors & Fragrances, Volatiles, Environmental
Featured Industry Environmental
Food and Beverages
suitability application for SPME, application for GC