GC Analysis of Water (0.01-0.5%Calibration Curve) on Watercol™ 1910
GC Analysis of Water (0.01-0.5%Calibration Curve) on Watercol™ 1910
column Watercol 1910, 30 m x 0.25 mm I.D., 0.20 µm (29711-U)
oven 80 °C (10 min)
carrier gas helium, 26 cm/sec
sample 5 standards, water at 0.01, 0.05, 0.1, 0.3, and 0.5% (v/v), each in ethanol
injection 0.5 µL, 100:1 split
liner 4 mm I.D., split type, cup design
inj. temp. 250 °C
detector TCD, 200 °C
Analysis Note The "Watercol" series of ionic liquid capillary GC columns are characterized by their ability to produce a sharp peak shape for water, sharp enough so that water can be integrated and quantified. This data shows the linearity obtained from a 5-point calibration curve (0.01-0.5%) for water on Watercol 1910.
Featured Industry Chemicals and Industrial Polymers
Cosmetics, Personal Care, and Cleaning Products
Flavors and Fragrances
Food and Beverages
Pharmaceutical (small molecule)
Legal Information Watercol is a trademark of Sigma-Aldrich Co. LLC