GC Analysis of Water (0.1-100%Calibration Curve) on Watercol™ 1460
GC Analysis of Water (0.1-100%Calibration Curve) on Watercol™ 1460
column Watercol 1460, 30 m x 0.25 mm I.D., 0.20 µm (29695-U)
oven 60 °C (12 min)
carrier gas helium, 26 cm/sec
sample 7 standards, water at 0.1, 0.5, 20, 40, 60, and 80% (v/v), each in ethanol, and water at 100%
injection 0.5 µL for 0.1-60% water, 0.1 µL for 80-100% water, 100:1 split
liner 4 mm I.D., split type, cup design
inj. temp. 250 °C
detector TCD, 200 °C
Analysis Note The "Watercol" series of ionic liquid capillary GC columns are characterized by their ability to produce a sharp peak shape for water, sharp enough so that water can be integrated and quantified. This data shows the linearity obtained from a 7-point calibration curve (0.1-100%) for water on Watercol 1460.
Featured Industry Chemicals and Industrial Polymers
Cosmetics, Personal Care, and Cleaning Products
Flavors and Fragrances
Food and Beverages
Pharmaceutical (small molecule)
Legal Information Watercol is a trademark of Sigma-Aldrich Co. LLC