GC Analysis of Water in Acetone (0.5 mg/mL Standard) on Watercol™ 1910
GC Analysis of Water in Acetone (0.5 mg/mL Standard) on Watercol™ 1910
Conditions
column Watercol 1910, 30 m x 0.25 mm I.D., 0.20 µm (29711-U)
oven 35 °C (4 min), 8 °C/min to 130 °C (1 min)
carrier gas helium, 11.40 psi constant pressure (equal to 25 cm/sec), measured with oven at 35 °C
sample 0.5 mg deionized water weighed into a 1 mL volumetric flask, acetone added to the 1 mL line, vortexed, transferred to an autosampler vial
injection 1 µL, 100:1 split
liner 4 mm I.D., split/splitless type, wool packed single taper FocusLiner™ design (2879925-U)
inj. temp. 250 °C
detector TCD, 200
Description
Analysis Note The "Watercol" series of ionic liquid capillary GC columns are characterized by their ability to produce a sharp peak shape for water, sharp enough so that water can be integrated and quantified. This application shows a calibration standard (0.5 mg/mL water in acetone) on Watercol 1910.
Featured Industry Chemicals and Industrial Polymers
Cosmetics, Personal Care, and Cleaning Products
Flavors and Fragrances
Food and Beverages
Petroleum
Pharmaceutical (small molecule)
Legal Information Watercol is a trademark of Sigma-Aldrich Co. LLC