Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Water in Ethanol (0.5%Standard) on Watercol™ 1460
GC Analysis of Water in Ethanol (0.5%Standard) on Watercol™ 1460
Conditions
column Watercol 1460, 30 m x 0.25 mm I.D., 0.20 µm (29695-U)
oven 60 °C (12 min)
carrier gas helium, 26 cm/sec
sample water at 0.05% (v/v) in ethanol
injection 0.5 µL, 100:1 split
liner 4 mm I.D., split type, cup design
inj. temp. 250 °C
detector TCD, 200 °C
Description
Analysis Note The "Watercol" series of ionic liquid capillary GC columns are characterized by their ability to produce a sharp peak shape for water, sharp enough so that water can be integrated and quantified. This application shows a calibration standard (0.5% water in ethanol) on Watercol 1460.
Featured Industry Chemicals and Industrial Polymers
Cosmetics, Personal Care, and Cleaning Products
Flavors and Fragrances
Food and Beverages
Petroleum
Pharmaceutical (small molecule)
Legal Information Watercol is a trademark of Sigma-Aldrich Co. LLC