Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Xylene Isomers on SLB®-IL60
GC Analysis of Xylene Isomers on SLB®-IL60
Conditions
column SLB-IL60, 30 m x 0.25 mm I.D., 0.20 µm (29505-U)
oven 40 °C (4 min), 8 °C/min to 200 °C (5 min)
carrier gas 30 cm/sec constant pressure
sample 12 analytes, each at 0.2% (v/v) in pentane
injection 1 µL100:1 split
liner 4 mm I.D., split/splitless type, wool packed single taper FocusLiner™ design
inj. temp. 250 °C
detector FID, 250 °C
Description
Analysis Note The xylene isomers are precursors to many chemicals:
• o-xylene is a precursor for phthalic anhydride
• m-xylene is a precursor for isophthalic acid
• p-xylene is a precursor for terephthalic acid and dimethyl terephthalate
The cresol (methylphenol) isomers are also precursors to many chemicals. This chromatogram of a mix of aromatic and methylphenol compounds was generated using an SLB-IL60 ionic liquid column. Its interaction mechanisms allow the separation of all three xylene isomers, and all three cresol isomers.
Categories Analytical Chromatography, GC Applications, Aromatics, Hydrocarbons
Featured Industry Chemicals and Industrial Polymers
Legal Information FocusLiner is a trademark of SGE Analytical Science Pty Ltd., SLB is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for GC
Analyte
  • 1,4-Dichlorobenzene
  • 2-Methylphenol
  • 3-Methylphenol
  • 4-Methylphenol
  • Benzene
  • Chlorobenzene
  • Ethylbenzene
  • m-Xylene
  • o-Xylene
  • p-Xylene
  • Styrene
  • Toluene