Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Xylenols on SLB(R)-IL76i
GC Analysis of Xylenols on SLB®-IL76i
Conditions
column SLB®-IL76i, 30 m x 0.25 mm I.D., 0.20 μm (29869-U)
oven 150 °C
carrier gas helium, 30 cm/sec, constant
sample 6 xylenols, each at 100 μg/mL in dichloromethane
injection 1 μL, 100:1 split
liner 3.5 mm I.D., splitless type, wool packed
inj. temp. 250 °C
detector MSD, m/z 50-500
MSD interface 270 °C
Description
Analysis Note Xylenols can be used as flavor ingredients in the food and beverage industry, or as industrial synthetic precursors for small molecule and polymer synthesis.
This application demonstrates the suitability of the SLB®-IL76i stationary phase for phenolic compounds, particularly xylenols (dimethylphenols). The ionic liquid stationary phase provides good retention and selectivity of these isomeric compounds. The inert nature of the deactivated column surface limits secondary interactions, yielding excellent peak shape for compounds with active moeities or the potential to hydrogen bond.
Featured Industry Chemicals and Industrial Polymers
Flavors and Fragrances
Food and Beverages
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC