GC/BID Analysis of Aromatics and Water in Solvents on Watercol™ 1910
GC/BID Analysis of Aromatics and Water in Solvents on Watercol 1910
Conditions
column Watercol™ 1910   30m x 0.25mm x 0.20 µm (29711-U)
oven 40°C (10 minutes) 8°C/minute to 170°C (5 minutes)
carrier gas helium, 25 cm/sec
sample BTEX spiked with 20ul of di-water in Hexane
injection 1 µL, 100:1 split
liner Split with wool
inj. temp. 250 °C
detector BID, 200 °C
Description
Analysis Note Easily determine water content in solvents with high aromatic content with one easy GC method. It is very common to  screen for aromatic impurities and moisture in solvents, but two different methods generally need to occur. Most commonly, a Karl Fischer method for water determination, as well as a GC method for aromatic impurities that can take up to an hour analysis time. With a combination of the Watercol 1910 column from MilliporeSigma, and the GC/BID from Shimadzu, this can be done in less that 20 minutes with one column and fantastic results. The BID shows a great response for aromatics and water with a stable baseline.
Featured Industry Petroleum 
Legal Information Watercol is a trademark of Sigma-Aldrich Co. LLC
Supelco is a trademark of Sigma-Aldrich Co. LLC
Application No. MCC310118