Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on SLB®-5ms
US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on SLB®-5ms
Conditions
column SLB-5ms, 30 m x 0.25 mm I.D., 0.25 µm (28471-U)
oven 100 °C (2 min), 15 °C/min to 300 °C (5 min)
carrier gas helium, 0.9 mL/min, constant flow
sample chlorinated pesticide standard (46845-U), diluted to 50 ppb in n-hexane
injection 2.0 µL, splitless (0.75 min)
liner 4 mm I.D., single taper
inj. temp. 250 °C
detector ECD, 300 °C
Description
Categories Analytical Chromatography, GC Applications, Pesticides
Featured Industry Environmental
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 1,1,1-Trichloro-2,2-bis(4-chlorophenyl)ethane (4,4′-DDT)
  • 1,1-Dichloro-2,2-bis(4-chlorophenyl)ethane (4,4′-DDD)
  • 1,1-Dichloro-2,2-bis(4-chlorophenyl)ethene
  • 2,4,5,6-Tetrachloro-m-xylene
  • Aldrin
  • Decachlorobiphenyl
  • Dieldrin
  • Endosulfan I
  • Endosulfan II
  • Endosulfan sulfate
  • Endrin
  • Endrin aldehyde
  • Endrin ketone
  • Heptachlor
  • Heptachlor epoxide
  • Methoxychlor
  • α-BHC
  • α-Chlordane
  • β-BHC
  • γ-BHC
  • γ-Chlordane
  • δ-BHC