Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on SLB®-5ms, 340 °C Final Temp.
US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on SLB®-5ms, 340 °C Final Temp.
Conditions
column SLB-5ms, 30 m x 0.25 mm I.D., 0.25 µm (28471-U)
oven 100 °C (2 min), 15 °C/min to 340 °C (5 min)
carrier gas helium, 0.9 mL/min
sample 200 ppm organochlorine pesticide standard mix (CRM46845) containing 22 analytes, diluted to 50 ppb with hexane
injection 1µL, splitless (0.75 min)
liner 4 mm I.D., split/splitless type, wool packed single taper FocusLiner™ design
inj. temp. 250 °C
detector muECD, 340 °C
Description
Analysis Note The analysis of low levels of 20 organochlorine pesticides and 2 surrogate compounds by gas chromatography-electron capture detector (GC-ECD) is routinely performed by analysts in environmental laboratories. Methods require the analysis of each sample extract on two columns with differing selectivity for confirmation of identifications. The SLB-5ms/SLB-35ms pair is an effective column set because all 22 analytes are resolved between both columns using the same run conditions.
This applicatin shows the separation achieved on SLB-5ms. Peaks are well integrated and baselines are exceptionally stable considering the high final oven temperature (340 °C). Minimal baseline rise is detected, and the last analyte, decachlorobiphenyl, is eluted in less than 20 minutes.
Categories Analytical Chromatography, GC Applications, Pesticides
Featured Industry Environmental
Legal Information FocusLiner is a trademark of SGE Analytical Science Pty Ltd., SLB is a registered trademark of Sigma-Aldrich Co. LLC