Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
US EPA Method 8015 (modified): GC Analysis of Gasoline Range Organics (GRO) on Equity®-1 (15 m x 0.10 mm I.D., 0.10 μm), Direct Injection, Fast GC Analysis
US EPA Method 8015 (modified): GC Analysis of Gasoline Range Organics (GRO) on Equity®-1 (15 m x 0.10 mm I.D., 0.10 μm), Direct Injection, Fast GC Analysis
Conditions
column Equity-1, 15 m x 0.10 mm I.D., 0.10 µm (28039-U)
oven 75 °C, 40 °C/min to 110 °C, 7.5 °C/min to 190 °C
carrier gas hydrogen, 57 cm/sec @ 75 °C
sample UST Modified GRO Mix, each analyte at 1000 ppm in methanol
injection 0.5 µL, 200:1 split
liner 4 mm I.D., split, cup design
inj. temp. 200 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Hydrocarbons
Featured Industry Environmental
Legal Information Equity is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 1,2,4-Trimethylbenzene
  • Benzene
  • Ethylbenzene
  • Mesitylene
  • m-Xylene
  • Naphthalene
  • o-Xylene
  • p-Xylene
  • tert-Butyl methyl ether
  • Toluene