Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
US EPA Method 8082: GC Analysis of Aroclor 1016/1260 (PCBs) on SLB®-5ms (20 m x 0.18 mm I.D., 0.36 μm), Fast GC Analysis
US EPA Method 8082: GC Analysis of Aroclor 1016/1260 (PCBs) on SLB®-5ms (20 m x 0.18 mm I.D., 0.36 μm), Fast GC Analysis
Conditions
column SLB-5ms, 20 m x 0.18 mm I.D., 0.36 µm (28576-U)
oven 100 °C (2 min), 15 °C/min to 325 °C (3 min)
carrier gas helium, 0.5 mL/min, constant flow
sample Aroclor standard mix 1 (46846-U) diluted to 500 ppb / 50 ppb (Aroclors / surrogates) in n-hexane
injection 1.0 µL, splitless (0.75 min)
liner 4 mm I.D., single taper
inj. temp. 250 °C
detector micro-ECD, 325 °C
Description
Categories Analytical Chromatography, GC Applications, Dioxins/PCBs/PBDEs
Featured Industry Environmental
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes