Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
US EPA Method 8082: GC Analysis of Aroclor 1016/1260 (PCBs) on SLB®-5ms (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
US EPA Method 8082: GC Analysis of Aroclor 1016/1260 (PCBs) on SLB®-5ms (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Conditions
column SLB-5ms, 15 m x 0.10 mm I.D., 0.10 µm (28466-U)
oven 80 °C (0.5 min), 50 °C/min to 200 °C, 35 °C/min to 360 °C (2 min)
carrier gas hydrogen, 40 cm/sec constant
sample Aroclor standard mix 1 (46846-U) diluted to 500 ppb / 50 ppb (Aroclors / surrogates) in n-hexane
injection 2 µL, splitless (0.75 min)
liner 4 mm I.D., single taper
inj. temp. 225 °C
detector ECD, 360 °C
Description
Categories Analytical Chromatography, GC Applications, Dioxins/PCBs/PBDEs
Featured Industry Environmental
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 2,4,5,6-Tetrachloro-m-xylene
  • Aroclor 1016
  • Aroclor 1260
  • Decachlorobiphenyl